The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002115.2 | NP_002106.2 | 17 - 18 | 2489 | 84 |
XM_017009411.1 | XP_016864900.1 | 16 - 17 | 2705 | 84 | |
GenBank mRNA | AB208951.1 | - | 14 - 15 | 3470 | 84 |
BC028129.1 | - | 17 - 18 | 2489 | 84 | |
U42303.1 | - | 8 - 9 | 1408 | 84 | |
U51333.1 | - | 17 - 18 | 2471 | 84 |