The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002115.2 | NP_002106.2 | 2 - 3 | 187 | 99 |
XM_011534540.2 | XP_011532842.1 | 2 - 3 | 228 | 99 | |
GenBank mRNA | AB208951.1 | - | 2 - 3 | 386 | 99 |
BC028129.1 | - | 2 - 3 | 187 | 99 | |
U51333.1 | - | 2 - 3 | 169 | 99 |