Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPA1L2
Entrez Gene ID
144983
Gene Name
heterogeneous nuclear ribonucleoprotein A1-like 2
Gene Aliases
-
Chromosome Location
Chr.13: 52617470 - 52643784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK123081.1 - 1 - 2 600 118

View Full Product Details