The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321291.1 | NP_001308220.1 | 9 - 10 | 1208 | 63 |
XM_005257542.2 | XP_005257599.1 | 9 - 10 | 1232 | 63 | |
XM_005257543.2 | XP_005257600.1 | 9 - 10 | 1232 | 63 | |
XM_011525065.1 | XP_011523367.1 | 9 - 10 | 1243 | 63 | |
XM_011525066.2 | XP_011523368.1 | 9 - 10 | 1105 | 63 | |
XM_011525067.2 | XP_011523369.1 | 9 - 10 | 1105 | 63 | |
XM_011525068.1 | XP_011523370.1 | 9 - 10 | 1146 | 63 | |
XM_011525069.2 | XP_011523371.1 | 9 - 10 | 1105 | 63 | |
XM_011525070.1 | XP_011523372.1 | 10 - 11 | 1176 | 63 | |
XM_011525071.2 | XP_011523373.1 | 9 - 10 | 1105 | 63 | |
XM_017024900.1 | XP_016880389.1 | 9 - 10 | 1232 | 63 | |
XM_017024901.1 | XP_016880390.1 | 7 - 8 | 767 | 63 | |
GenBank mRNA | AB040886.1 | - | 9 - 10 | 1125 | 63 |
AK001671.1 | - | 9 - 10 | 1243 | 63 | |
AK022840.1 | - | 3 - 4 | 1463 | 63 | |
AK022913.1 | - | 9 - 10 | 1150 | 63 | |
AK225426.1 | - | 9 - 10 | 1107 | 63 | |
AY169386.1 | - | 7 - 8 | 907 | 63 | |
BC016487.1 | - | 9 - 10 | 1209 | 63 | |
BC027992.1 | - | 9 - 10 | 1146 | 63 | |
BC071582.1 | - | 9 - 10 | 1208 | 63 |