The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303237.1 | NP_001290166.1 | 1 - 2 | 333 | 122 |
NM_002201.5 | NP_002192.2 | 1 - 2 | 333 | 122 | |
XM_017022147.1 | XP_016877636.1 | 1 - 2 | 1522 | 122 | |
GenBank mRNA | AK307769.1 | - | 1 - 2 | 136 | 122 |
BC007922.2 | - | 1 - 2 | 268 | 122 | |
BG759382.1 | - | 1 - 2 | 334 | 122 | |
BX339191.2 | - | 1 - 2 | 272 | 122 | |
CN265233.1 | - | 1 - 2 | 123 | 122 |