Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NUCB2
- Entrez Gene ID
-
4925
- Gene Name
- nucleobindin 2
- Gene Aliases
-
HEL-S-109, NEFA
- Chromosome Location
-
Chr.11: 17260340 - 17331881 on Build GRCh38
View Full Product Details