Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUCB2
Entrez Gene ID
4925
Gene Name
nucleobindin 2
Gene Aliases
HEL-S-109, NEFA
Chromosome Location
Chr.11: 17260340 - 17331881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017017809.1 XP_016873298.1 1 - 2 129 141
XM_017017814.1 XP_016873303.1 1 - 2 101 141
XM_017017826.1 XP_016873315.1 1 - 2 129 141
XM_017017828.1 XP_016873317.1 1 - 2 101 141
GenBank mRNA EU039832.1 - 1 - 2 113 141

View Full Product Details