The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005662.6 | NP_005653.3 | 4 - 5 | 328 | 102 |
GenBank mRNA | AF038962.1 | - | 4 - 5 | 217 | 102 |
AK058201.1 | - | 4 - 5 | 202 | 102 | |
AK307224.1 | - | 4 - 5 | 208 | 102 | |
BC056870.1 | - | 4 - 5 | 213 | 102 | |
DA930673.1 | - | 4 - 5 | 328 | 102 | |
U90943.1 | - | 2 - 3 | 118 | 102 |