Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 14 - 15 2867 82
XM_011527173.2 XP_011525475.1 14 - 15 2933 82
XM_017027049.1 XP_016882538.1 14 - 15 3120 82
XM_017027050.1 XP_016882539.1 12 - 13 2502 82
XM_017027051.1 XP_016882540.1 14 - 15 2933 82
XM_017027052.1 XP_016882541.1 14 - 15 2933 82
GenBank mRNA AB046862.1 - 5 - 6 1610 82
AF082075.1 - 14 - 15 2928 82
AF311855.1 - 14 - 15 2867 82
AF324063.1 - 14 - 15 2851 82
AF324064.1 - 14 - 15 2851 82
AY004226.1 - 13 - 14 2750 82
AY004227.1 - 13 - 14 2750 82

View Full Product Details