Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 20 - 21 4462 78
NM_025213.2 NP_079489.2 3 - 4 515 78
XM_011527173.2 XP_011525475.1 20 - 21 4528 78
XM_017027049.1 XP_016882538.1 20 - 21 4715 78
XM_017027050.1 XP_016882539.1 18 - 19 4097 78
XM_017027051.1 XP_016882540.1 20 - 21 4528 78
GenBank mRNA AB046862.1 - 11 - 12 3205 78
AF082075.1 - 20 - 21 4523 78
AF311855.1 - 20 - 21 4462 78
AF311856.1 - 3 - 4 515 78
AF324063.1 - 20 - 21 4446 78
AF324064.1 - 20 - 21 4446 78
AK309418.1 - 3 - 4 477 78
AY004226.1 - 20 - 21 4431 78
AY004227.1 - 19 - 20 4345 78
DA514440.1 - 3 - 4 477 78

View Full Product Details