The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020971.2 | NP_066022.2 | 8 - 9 | 1000 | 137 |
XM_011527173.2 | XP_011525475.1 | 8 - 9 | 1066 | 137 | |
XM_017027049.1 | XP_016882538.1 | 8 - 9 | 1253 | 137 | |
XM_017027050.1 | XP_016882539.1 | 6 - 7 | 635 | 137 | |
XM_017027051.1 | XP_016882540.1 | 8 - 9 | 1066 | 137 | |
XM_017027052.1 | XP_016882541.1 | 8 - 9 | 1066 | 137 | |
GenBank mRNA | AF082075.1 | - | 8 - 9 | 1076 | 137 |
AF311855.1 | - | 8 - 9 | 1000 | 137 | |
AF324063.1 | - | 8 - 9 | 984 | 137 | |
AF324064.1 | - | 8 - 9 | 984 | 137 | |
AY004226.1 | - | 7 - 8 | 898 | 137 | |
AY004227.1 | - | 7 - 8 | 898 | 137 |