Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 9 - 10 1182 67
XM_011527173.2 XP_011525475.1 9 - 10 1248 67
XM_017027049.1 XP_016882538.1 9 - 10 1435 67
XM_017027050.1 XP_016882539.1 7 - 8 817 67
XM_017027051.1 XP_016882540.1 9 - 10 1248 67
XM_017027052.1 XP_016882541.1 9 - 10 1248 67
GenBank mRNA AF082075.1 - 9 - 10 1258 67
AF311855.1 - 9 - 10 1182 67
AF324063.1 - 9 - 10 1166 67
AF324064.1 - 9 - 10 1166 67
AY004226.1 - 8 - 9 1080 67
AY004227.1 - 8 - 9 1080 67

View Full Product Details