Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSBPL10
Entrez Gene ID
114884
Gene Name
oxysterol binding protein like 10
Gene Aliases
ORP10, OSBP9
Chromosome Location
Chr.3: 31660825 - 31981850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF392451.1 - 1 - 2 256 55

View Full Product Details