The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081560.2 | NP_001075029.1 | 11 - 12 | 1696 | 78 |
NM_001081562.2 | NP_001075031.1 | 11 - 12 | 1696 | 78 | |
NM_001081563.2 | NP_001075032.1 | 10 - 11 | 2080 | 78 | |
NM_001288764.1 | NP_001275693.1 | 12 - 13 | 1770 | 78 | |
NM_001288765.1 | NP_001275694.1 | 9 - 10 | 1351 | 78 | |
NM_001288766.1 | NP_001275695.1 | 11 - 12 | 1696 | 78 | |
NM_004409.4 | NP_004400.4 | 11 - 12 | 1711 | 78 | |
GenBank mRNA | AB209363.1 | - | 11 - 12 | 2539 | 78 |
AK297299.1 | - | 9 - 10 | 1302 | 78 | |
AK302874.1 | - | 11 - 12 | 1629 | 78 | |
BC026328.1 | - | 12 - 13 | 1765 | 78 | |
BC062553.1 | - | 11 - 12 | 1632 | 78 | |
BP422066.1 | - | 1 - 2 | 41 | 78 | |
KJ534826.1 | - | 10 - 11 | 1536 | 78 | |
KJ534827.1 | - | 10 - 11 | 1536 | 78 | |
KJ534828.1 | - | 10 - 11 | 1521 | 78 | |
L19268.1 | - | 11 - 12 | 1618 | 78 | |
M87312.1 | - | 10 - 11 | 2081 | 78 | |
M94203.1 | - | 11 - 12 | 1368 | 78 | |
S72883.1 | - | 11 - 12 | 1527 | 78 |