The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081560.2 | NP_001075029.1 | 4 - 5 | 634 | 89 |
NM_001081562.2 | NP_001075031.1 | 4 - 5 | 634 | 89 | |
NM_001081563.2 | NP_001075032.1 | 3 - 4 | 1003 | 89 | |
NM_001288764.1 | NP_001275693.1 | 5 - 6 | 693 | 89 | |
NM_001288765.1 | NP_001275694.1 | 2 - 3 | 274 | 89 | |
NM_001288766.1 | NP_001275695.1 | 4 - 5 | 634 | 89 | |
NM_004409.4 | NP_004400.4 | 4 - 5 | 634 | 89 | |
GenBank mRNA | AB209363.1 | - | 4 - 5 | 1477 | 89 |
AK297299.1 | - | 2 - 3 | 225 | 89 | |
AK302874.1 | - | 4 - 5 | 567 | 89 | |
BC026328.1 | - | 5 - 6 | 688 | 89 | |
BC062553.1 | - | 4 - 5 | 555 | 89 | |
DC420602.1 | - | 2 - 3 | 257 | 89 | |
KJ534826.1 | - | 3 - 4 | 459 | 89 | |
KJ534827.1 | - | 3 - 4 | 459 | 89 | |
KJ534828.1 | - | 3 - 4 | 459 | 89 | |
L19268.1 | - | 4 - 5 | 556 | 89 | |
M87312.1 | - | 3 - 4 | 1004 | 89 | |
M94203.1 | - | 4 - 5 | 306 | 89 | |
S72883.1 | - | 4 - 5 | 465 | 89 |