The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 13 - 14 | 1776 | 70 |
NM_015386.2 | NP_056201.2 | 14 - 15 | 1839 | 70 | |
XM_011522981.2 | XP_011521283.1 | 15 - 16 | 1999 | 70 | |
GenBank mRNA | AB088369.1 | - | 15 - 16 | 1975 | 70 |
AK022874.1 | - | 14 - 15 | 1819 | 70 | |
AK026435.1 | - | 10 - 11 | 1133 | 70 | |
AK096557.1 | - | 12 - 13 | 1557 | 70 | |
AK293235.1 | - | 11 - 12 | 1383 | 70 | |
AK297557.1 | - | 15 - 16 | 1782 | 70 | |
AL050101.1 | - | 10 - 11 | 1141 | 70 | |
BC006306.2 | - | 11 - 12 | 1355 | 70 | |
BC013347.2 | - | 10 - 11 | 1147 | 70 | |
BC027726.2 | - | 13 - 14 | 1750 | 70 | |
BC072438.1 | - | 14 - 15 | 1819 | 70 | |
BX648010.1 | - | 12 - 13 | 2507 | 70 |