Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location
Chr.17: 16030094 - 16216746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190438.1 NP_001177367.1 7 - 8 821 137
NM_001190440.1 NP_001177369.1 9 - 10 1148 137
NM_006311.3 NP_006302.2 10 - 11 1347 137
XM_005256866.4 XP_005256923.1 11 - 12 1357 137
XM_005256868.4 XP_005256925.1 11 - 12 1357 137
XM_005256871.4 XP_005256928.1 10 - 11 1330 137
XM_005256872.4 XP_005256929.1 10 - 11 1330 137
XM_005256873.4 XP_005256930.1 11 - 12 1357 137
XM_005256874.4 XP_005256931.1 11 - 12 1357 137
XM_005256875.3 XP_005256932.1 11 - 12 1357 137
XM_006721601.3 XP_006721664.1 12 - 13 1489 137
XM_006721602.3 XP_006721665.1 12 - 13 1487 137
XM_006721603.3 XP_006721666.1 11 - 12 1357 137
XM_006721604.3 XP_006721667.1 10 - 11 1330 137
XM_011524083.2 XP_011522385.1 11 - 12 2268 137
XM_011524084.2 XP_011522386.1 11 - 12 1357 137
XM_011524085.2 XP_011522387.1 11 - 12 1357 137
XM_011524086.2 XP_011522388.1 10 - 11 1330 137
XM_017025396.1 XP_016880885.1 12 - 13 2234 137
XM_017025397.1 XP_016880886.1 12 - 13 1999 137
XM_017025398.1 XP_016880887.1 11 - 12 1441 137
XM_017025399.1 XP_016880888.1 11 - 12 1460 137
XM_017025400.1 XP_016880889.1 12 - 13 2234 137
XM_017025401.1 XP_016880890.1 12 - 13 1999 137
XM_017025402.1 XP_016880891.1 11 - 12 1283 137
XM_017025403.1 XP_016880892.1 11 - 12 1457 137
XM_017025404.1 XP_016880893.1 11 - 12 1948 137
XM_017025405.1 XP_016880894.1 10 - 11 1843 137
XM_017025406.1 XP_016880895.1 10 - 11 2241 137
XM_017025407.1 XP_016880896.1 10 - 11 1177 137
XM_017025408.1 XP_016880897.1 10 - 11 1249 137
XM_017025409.1 XP_016880898.1 11 - 12 1457 137
XM_017025410.1 XP_016880899.1 11 - 12 1953 137
XM_017025411.1 XP_016880900.1 10 - 11 1696 137
XM_017025412.1 XP_016880901.1 10 - 11 1473 137
XM_017025413.1 XP_016880902.1 10 - 11 1256 137
XM_017025414.1 XP_016880903.1 9 - 10 1163 137
XM_017025415.1 XP_016880904.1 11 - 12 1357 137
XM_017025416.1 XP_016880905.1 11 - 12 1968 137
XM_017025417.1 XP_016880906.1 11 - 12 1357 137
XM_017025418.1 XP_016880907.1 11 - 12 1357 137
XM_017025419.1 XP_016880908.1 11 - 12 1357 137
XM_017025420.1 XP_016880909.1 11 - 12 1357 137
GenBank mRNA AB028970.2 - 9 - 10 1122 137
AF044209.1 - 10 - 11 1318 137
AF087856.1 - 7 - 8 835 137
AF303586.1 - 7 - 8 835 137
AK307614.1 - 11 - 12 1343 137
BC026028.1 - 12 - 13 1448 137
BC050594.2 - 10 - 11 1319 137
BC056862.1 - 10 - 11 1263 137
BC058511.1 - 10 - 11 1324 137
BC068996.1 - 10 - 11 1319 137
BC142648.1 - 10 - 11 1301 137
BC167431.1 - 9 - 10 1123 137

View Full Product Details