The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190440.1 | NP_001177369.1 | 21 - 22 | 3138 | 77 |
NM_006311.3 | NP_006302.2 | 22 - 23 | 3289 | 77 | |
XM_005256874.4 | XP_005256931.1 | 23 - 24 | 3350 | 77 | |
XM_006721604.3 | XP_006721667.1 | 22 - 23 | 3320 | 77 | |
XM_011524086.2 | XP_011522388.1 | 22 - 23 | 3320 | 77 | |
XM_017025415.1 | XP_016880904.1 | 23 - 24 | 3350 | 77 | |
XM_017025416.1 | XP_016880905.1 | 23 - 24 | 3958 | 77 | |
XM_017025417.1 | XP_016880906.1 | 23 - 24 | 3302 | 77 | |
XM_017025419.1 | XP_016880908.1 | 23 - 24 | 3350 | 77 | |
XM_017025420.1 | XP_016880909.1 | 23 - 24 | 3302 | 77 | |
GenBank mRNA | AB028970.2 | - | 21 - 22 | 3112 | 77 |
AF087856.1 | - | 19 - 20 | 2825 | 77 | |
BC167431.1 | - | 21 - 22 | 3113 | 77 |