Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location
Chr.17: 16030094 - 16216746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190440.1 NP_001177369.1 38 - 39 5926 82
NM_006311.3 NP_006302.2 39 - 40 6434 82
XM_005256866.4 XP_005256923.1 41 - 42 6669 82
XM_005256868.4 XP_005256925.1 41 - 42 6666 82
XM_005256871.4 XP_005256928.1 40 - 41 6642 82
XM_005256872.4 XP_005256929.1 40 - 41 6639 82
XM_005256873.4 XP_005256930.1 41 - 42 6621 82
XM_005256874.4 XP_005256931.1 40 - 41 6495 82
XM_005256875.3 XP_005256932.1 40 - 41 6441 82
XM_006721601.3 XP_006721664.1 42 - 43 6801 82
XM_006721602.3 XP_006721665.1 42 - 43 6796 82
XM_006721603.3 XP_006721666.1 41 - 42 6669 82
XM_006721604.3 XP_006721667.1 39 - 40 6465 82
XM_011524083.2 XP_011522385.1 41 - 42 7580 82
XM_011524084.2 XP_011522386.1 41 - 42 6669 82
XM_011524085.2 XP_011522387.1 41 - 42 6312 82
XM_011524086.2 XP_011522388.1 39 - 40 6108 82
XM_017025396.1 XP_016880885.1 42 - 43 7546 82
XM_017025397.1 XP_016880886.1 42 - 43 7311 82
XM_017025398.1 XP_016880887.1 41 - 42 6753 82
XM_017025399.1 XP_016880888.1 41 - 42 6772 82
XM_017025400.1 XP_016880889.1 42 - 43 7543 82
XM_017025401.1 XP_016880890.1 42 - 43 7308 82
XM_017025402.1 XP_016880891.1 41 - 42 6592 82
XM_017025403.1 XP_016880892.1 41 - 42 6769 82
XM_017025404.1 XP_016880893.1 41 - 42 7260 82
XM_017025405.1 XP_016880894.1 40 - 41 7155 82
XM_017025406.1 XP_016880895.1 40 - 41 7553 82
XM_017025407.1 XP_016880896.1 40 - 41 6489 82
XM_017025408.1 XP_016880897.1 40 - 41 6561 82
XM_017025409.1 XP_016880898.1 41 - 42 6766 82
XM_017025410.1 XP_016880899.1 41 - 42 7262 82
XM_017025411.1 XP_016880900.1 40 - 41 7005 82
XM_017025412.1 XP_016880901.1 40 - 41 6782 82
XM_017025413.1 XP_016880902.1 40 - 41 6565 82
XM_017025414.1 XP_016880903.1 39 - 40 6472 82
XM_017025415.1 XP_016880904.1 40 - 41 6495 82
XM_017025416.1 XP_016880905.1 40 - 41 7103 82
XM_017025417.1 XP_016880906.1 40 - 41 6447 82
XM_017025418.1 XP_016880907.1 41 - 42 6312 82
XM_017025419.1 XP_016880908.1 40 - 41 6138 82
XM_017025420.1 XP_016880909.1 40 - 41 6090 82
GenBank mRNA AB028970.2 - 38 - 39 5900 82
AF044209.1 - 39 - 40 6405 82
AF087856.1 - 36 - 37 5961 82
AF303584.1 - 1 - 2 414 82
AF303585.1 - 8 - 9 1940 82
AK295893.1 - 10 - 11 2057 82
AK310405.1 - 15 - 16 2937 82
AL137641.1 - 9 - 10 1987 82
BC167431.1 - 38 - 39 5901 82

View Full Product Details