Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location
Chr.17: 16030094 - 16216746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190440.1 NP_001177369.1 2 - 3 308 90
NM_006311.3 NP_006302.2 3 - 4 507 90
XM_005256866.4 XP_005256923.1 3 - 4 490 90
XM_005256868.4 XP_005256925.1 3 - 4 490 90
XM_005256871.4 XP_005256928.1 3 - 4 490 90
XM_005256872.4 XP_005256929.1 3 - 4 490 90
XM_005256873.4 XP_005256930.1 3 - 4 490 90
XM_005256874.4 XP_005256931.1 3 - 4 490 90
XM_005256875.3 XP_005256932.1 3 - 4 490 90
XM_006721601.3 XP_006721664.1 4 - 5 622 90
XM_006721602.3 XP_006721665.1 4 - 5 620 90
XM_006721603.3 XP_006721666.1 3 - 4 490 90
XM_006721604.3 XP_006721667.1 3 - 4 490 90
XM_011524083.2 XP_011522385.1 3 - 4 1401 90
XM_011524084.2 XP_011522386.1 3 - 4 490 90
XM_011524085.2 XP_011522387.1 3 - 4 490 90
XM_011524086.2 XP_011522388.1 3 - 4 490 90
XM_017025396.1 XP_016880885.1 4 - 5 1367 90
XM_017025397.1 XP_016880886.1 4 - 5 1132 90
XM_017025398.1 XP_016880887.1 3 - 4 574 90
XM_017025399.1 XP_016880888.1 3 - 4 593 90
XM_017025400.1 XP_016880889.1 4 - 5 1367 90
XM_017025401.1 XP_016880890.1 4 - 5 1132 90
XM_017025402.1 XP_016880891.1 3 - 4 416 90
XM_017025403.1 XP_016880892.1 4 - 5 617 90
XM_017025404.1 XP_016880893.1 4 - 5 1108 90
XM_017025405.1 XP_016880894.1 3 - 4 1003 90
XM_017025406.1 XP_016880895.1 3 - 4 1401 90
XM_017025407.1 XP_016880896.1 3 - 4 337 90
XM_017025408.1 XP_016880897.1 3 - 4 409 90
XM_017025409.1 XP_016880898.1 4 - 5 617 90
XM_017025410.1 XP_016880899.1 4 - 5 1113 90
XM_017025411.1 XP_016880900.1 3 - 4 856 90
XM_017025412.1 XP_016880901.1 3 - 4 633 90
XM_017025413.1 XP_016880902.1 3 - 4 416 90
XM_017025414.1 XP_016880903.1 2 - 3 323 90
XM_017025415.1 XP_016880904.1 3 - 4 490 90
XM_017025416.1 XP_016880905.1 4 - 5 1128 90
XM_017025417.1 XP_016880906.1 3 - 4 490 90
XM_017025418.1 XP_016880907.1 3 - 4 490 90
XM_017025419.1 XP_016880908.1 3 - 4 490 90
XM_017025420.1 XP_016880909.1 3 - 4 490 90
GenBank mRNA AB028970.2 - 2 - 3 282 90
AF044209.1 - 3 - 4 478 90
AK307614.1 - 3 - 4 476 90
BC026028.1 - 4 - 5 581 90
BC050594.2 - 3 - 4 479 90
BC056862.1 - 3 - 4 423 90
BC058511.1 - 3 - 4 484 90
BC068996.1 - 3 - 4 479 90
BC107772.1 - 3 - 4 495 90
BC142648.1 - 3 - 4 461 90
BC167431.1 - 2 - 3 283 90

View Full Product Details