Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKLE1
Entrez Gene ID
126549
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
ANKRD41, LEM3, LEMD6
Chromosome Location
Chr.19: 17281645 - 17287646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278444.1 NP_001265373.1 1 - 2 341 131
NM_152363.5 NP_689576.5 1 - 2 341 131
NR_103530.1 1 - 2 341 131
GenBank mRNA AK303628.1 - 1 - 2 341 131
DC410789.1 - 1 - 2 341 131

View Full Product Details