The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100412.1 | NP_001093882.1 | 25 - 26 | 3463 | 71 |
NM_013235.4 | NP_037367.3 | 25 - 26 | 3498 | 71 | |
XM_005248291.3 | XP_005248348.1 | 26 - 27 | 3574 | 71 | |
XM_005248292.3 | XP_005248349.1 | 26 - 27 | 3550 | 71 | |
XM_005248293.3 | XP_005248350.1 | 25 - 26 | 3481 | 71 | |
XM_005248294.3 | XP_005248351.1 | 24 - 25 | 3370 | 71 | |
XM_011514033.2 | XP_011512335.1 | 26 - 27 | 3574 | 71 | |
XM_017009399.1 | XP_016864888.1 | 25 - 26 | 3457 | 71 | |
XM_017009400.1 | XP_016864889.1 | 25 - 26 | 3439 | 71 | |
XM_017009401.1 | XP_016864890.1 | 24 - 25 | 3346 | 71 | |
XM_017009402.1 | XP_016864891.1 | 14 - 15 | 1589 | 71 | |
GenBank mRNA | AF116910.1 | - | 10 - 11 | 1027 | 71 |
AF189011.1 | - | 25 - 26 | 3384 | 71 | |
AK001121.1 | - | 14 - 15 | 1333 | 71 | |
AK026563.1 | - | 5 - 6 | 385 | 71 | |
AK308093.1 | - | 24 - 25 | 3272 | 71 | |
BC024261.1 | - | 13 - 14 | 1259 | 71 | |
BC041162.1 | - | 12 - 13 | 1184 | 71 | |
BC054003.1 | - | 24 - 25 | 3275 | 71 | |
BX647724.1 | - | 25 - 26 | 3366 | 71 |