The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002833.3 | NP_002824.1 | 7 - 8 | 1480 | 76 |
GenBank mRNA | BC010863.1 | - | 7 - 8 | 984 | 76 |
BC071574.1 | - | 8 - 9 | 1401 | 76 | |
BT007405.1 | - | 7 - 8 | 965 | 76 | |
M83738.1 | - | 7 - 8 | 1473 | 76 |