The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016327.2 | NP_057411.1 | 4 - 5 | 584 | 60 |
XM_011530222.2 | XP_011528524.1 | 6 - 7 | 888 | 60 | |
XM_011530223.2 | XP_011528525.1 | 6 - 7 | 888 | 60 | |
XM_011530224.2 | XP_011528526.1 | 6 - 7 | 888 | 60 | |
XM_011530225.2 | XP_011528527.1 | 5 - 6 | 584 | 60 | |
XM_017028825.1 | XP_016884314.1 | 6 - 7 | 888 | 60 | |
XM_017028826.1 | XP_016884315.1 | 6 - 7 | 888 | 60 | |
XM_017028827.1 | XP_016884316.1 | 6 - 7 | 888 | 60 | |
XM_017028828.1 | XP_016884317.1 | 6 - 7 | 888 | 60 | |
GenBank mRNA | AB013885.1 | - | 4 - 5 | 523 | 60 |
AF163312.1 | - | 4 - 5 | 524 | 60 | |
AK024277.1 | - | 4 - 5 | 1852 | 60 | |
AK314727.1 | - | 4 - 5 | 720 | 60 | |
BC033388.1 | - | 4 - 5 | 481 | 60 | |
BC131703.1 | - | 4 - 5 | 537 | 60 | |
BC139843.1 | - | 4 - 5 | 482 | 60 | |
CR456375.1 | - | 4 - 5 | 584 | 60 |