The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016327.2 | NP_057411.1 | 7 - 8 | 991 | 70 |
XM_011530222.2 | XP_011528524.1 | 9 - 10 | 1295 | 70 | |
XM_011530225.2 | XP_011528527.1 | 8 - 9 | 991 | 70 | |
XM_017028826.1 | XP_016884315.1 | 9 - 10 | 1295 | 70 | |
XM_017028827.1 | XP_016884316.1 | 8 - 9 | 1125 | 70 | |
GenBank mRNA | AB013885.1 | - | 7 - 8 | 930 | 70 |
AF163312.1 | - | 7 - 8 | 931 | 70 | |
AK024277.1 | - | 7 - 8 | 2259 | 70 | |
AK314727.1 | - | 7 - 8 | 1127 | 70 | |
BC033388.1 | - | 7 - 8 | 888 | 70 | |
BC131703.1 | - | 7 - 8 | 944 | 70 | |
BC139843.1 | - | 7 - 8 | 889 | 70 | |
CR456375.1 | - | 7 - 8 | 991 | 70 |