Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
Chromosome Location
Chr.13: 94437304 - 94550265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129889.2 NP_001123361.1 4 - 5 1286 96
NM_001322182.1 NP_001309111.1 3 - 4 508 96
NM_001322183.1 NP_001309112.1 4 - 5 562 96
NM_001322184.1 NP_001309113.1 5 - 6 862 96
NM_001322185.1 NP_001309114.1 4 - 5 808 96
NM_001322186.1 NP_001309115.1 6 - 7 1077 96
NM_001922.4 NP_001913.2 4 - 5 1286 96
XM_011521049.1 XP_011519351.1 6 - 7 1910 96
XM_017020401.1 XP_016875890.1 4 - 5 1289 96
GenBank mRNA AK293115.1 - 4 - 5 1069 96
BC028311.1 - 4 - 5 1133 96
D17547.1 - 4 - 5 1273 96
DQ902581.1 - 4 - 5 1151 96
L18967.1 - 4 - 5 1243 96
S69231.1 - 4 - 5 1208 96

View Full Product Details