Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL1
Entrez Gene ID
4234
Gene Name
methyltransferase like 1
Gene Aliases
C12orf1, TRM8, TRMT8, YDL201w
Chromosome Location
Chr.12: 57768568 - 57772249 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268873.2 XP_005268930.1 2 - 3 254 57

View Full Product Details