The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003406.1 | NP_001003406.1 | 14 - 15 | 2592 | 111 |
NM_021096.3 | NP_066919.2 | 15 - 16 | 2697 | 111 | |
XM_017029035.1 | XP_016884524.1 | 5 - 6 | 5967 | 111 | |
XM_017029036.1 | XP_016884525.1 | 5 - 6 | 5967 | 111 | |
XM_017029037.1 | XP_016884526.1 | 5 - 6 | 5968 | 111 | |
GenBank mRNA | AF129133.1 | - | 15 - 16 | 2697 | 111 |
AF142567.1 | - | 15 - 16 | 2697 | 111 | |
AF211189.1 | - | 14 - 15 | 2592 | 111 | |
AF393329.1 | - | 14 - 15 | 2592 | 111 |