The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003406.1 | NP_001003406.1 | 16 - 17 | 3264 | 118 |
NM_021096.3 | NP_066919.2 | 17 - 18 | 3369 | 118 | |
XM_017029035.1 | XP_016884524.1 | 7 - 8 | 6639 | 118 | |
XM_017029036.1 | XP_016884525.1 | 7 - 8 | 6639 | 118 | |
XM_017029037.1 | XP_016884526.1 | 7 - 8 | 6640 | 118 | |
GenBank mRNA | AF129133.1 | - | 17 - 18 | 3369 | 118 |
AF142567.1 | - | 17 - 18 | 3369 | 118 | |
AF211189.1 | - | 16 - 17 | 3264 | 118 | |
AF393329.1 | - | 16 - 17 | 3264 | 118 |