Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
Chromosome Location
Chr.12: 95943293 - 95968592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152435.2 NP_689648.2 5 - 6 925 57
GenBank mRNA AB075878.1 - 5 - 6 880 57
AK054617.1 - 5 - 6 925 57
AK122802.1 - 5 - 6 892 57
AK290828.1 - 5 - 6 880 57
BC029146.1 - 5 - 6 861 57

View Full Product Details