The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152435.2 | NP_689648.2 | 7 - 8 | 1133 | 64 |
GenBank mRNA | AB075878.1 | - | 7 - 8 | 1088 | 64 |
AK054617.1 | - | 7 - 8 | 1133 | 64 | |
AK290828.1 | - | 7 - 8 | 1088 | 64 | |
AL833144.1 | - | 6 - 7 | 841 | 64 | |
BC029146.1 | - | 7 - 8 | 1069 | 64 |