The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127460.2 | NP_001120932.1 | 10 - 11 | 1978 | 74 |
NM_001166279.1 | NP_001159751.1 | 10 - 11 | 1907 | 74 | |
NM_014871.4 | NP_055686.3 | 10 - 11 | 1907 | 74 | |
GenBank mRNA | AB014610.2 | - | 10 - 11 | 1804 | 74 |
AB107585.1 | - | 9 - 10 | 1601 | 74 | |
AK310822.1 | - | 10 - 11 | 1791 | 74 | |
BC024043.1 | - | 10 - 11 | 1820 | 74 | |
BC094885.1 | - | 10 - 11 | 1975 | 74 | |
BX648106.1 | - | 10 - 11 | 1895 | 74 |