The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127460.2 | NP_001120932.1 | 15 - 16 | 2663 | 121 |
NM_001166279.1 | NP_001159751.1 | 15 - 16 | 2589 | 121 | |
NM_014871.4 | NP_055686.3 | 15 - 16 | 2580 | 121 | |
GenBank mRNA | AB014610.2 | - | 15 - 16 | 2486 | 121 |
AB107585.1 | - | 14 - 15 | 2274 | 121 | |
AK096679.1 | - | 3 - 4 | 348 | 121 | |
BC024043.1 | - | 15 - 16 | 2493 | 121 | |
BC094885.1 | - | 15 - 16 | 2660 | 121 | |
BX648106.1 | - | 15 - 16 | 2565 | 121 |