Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A1
Entrez Gene ID
931
Gene Name
membrane spanning 4-domains A1
Gene Aliases
B1, Bp35, CD20, CVID5, LEU-16, MS4A2, S7
Chromosome Location
Chr.11: 60455809 - 60470752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021950.3 NP_068769.2 3 - 4 435 68
NM_152866.2 NP_690605.1 4 - 5 698 68
GenBank mRNA AK225630.1 - 3 - 4 397 68
AK292168.1 - 3 - 4 571 68
BC002807.2 - 3 - 4 385 68
CD691924.1 - 4 - 5 458 68
X07203.1 - 3 - 4 372 68
X12530.1 - 3 - 4 375 68

View Full Product Details