Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADSYN1
Entrez Gene ID
55191
Gene Name
NAD synthetase 1
Gene Aliases
-
Chromosome Location
Chr.11: 71453171 - 71501538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018161.4 NP_060631.2 12 - 13 1170 72
GenBank mRNA AB091316.1 - 12 - 13 1044 72
AK001493.1 - 12 - 13 1111 72
AK022436.1 - 12 - 13 1140 72
AK094022.1 - 3 - 4 1152 72
AK097946.1 - 4 - 5 1049 72
BC003638.2 - 12 - 13 1133 72
BC003666.2 - 12 - 13 1133 72
CR457286.1 - 12 - 13 1044 72

View Full Product Details