Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IMPG2
Entrez Gene ID
50939
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, RP56, SPACRCAN, VMD5
Chromosome Location
Chr.3: 101222546 - 101320575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016247.3 NP_057331.2 12 - 13 1745 110
GenBank mRNA AB593127.1 - 12 - 13 1736 110
AF157624.1 - 12 - 13 1556 110
AF173155.1 - 12 - 13 1733 110

View Full Product Details