The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016247.3 | NP_057331.2 | 1 - 2 | 287 | 135 |
GenBank mRNA | AB593127.1 | - | 1 - 2 | 278 | 135 |
AF173155.1 | - | 1 - 2 | 275 | 135 | |
BX953286.1 | - | 1 - 2 | 288 | 135 |