The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080448.2 | NP_001073917.2 | 15 - 16 | 2978 | 93 |
XM_006713592.3 | XP_006713655.1 | 16 - 17 | 3026 | 93 | |
XM_017006210.1 | XP_016861699.1 | 15 - 16 | 2900 | 93 | |
XM_017006211.1 | XP_016861700.1 | 17 - 18 | 3051 | 93 | |
XM_017006212.1 | XP_016861701.1 | 16 - 17 | 2720 | 93 | |
XM_017006213.1 | XP_016861702.1 | 16 - 17 | 3078 | 93 | |
XM_017006214.1 | XP_016861703.1 | 15 - 16 | 2346 | 93 | |
XM_017006215.1 | XP_016861704.1 | 11 - 12 | 1375 | 93 | |
XM_017006216.1 | XP_016861705.1 | 10 - 11 | 1325 | 93 | |
XM_017006217.1 | XP_016861706.1 | 12 - 13 | 1422 | 93 | |
XM_017006218.1 | XP_016861707.1 | 10 - 11 | 1225 | 93 | |
XM_017006220.1 | XP_016861709.1 | 12 - 13 | 1390 | 93 | |
XM_017006221.1 | XP_016861710.1 | 10 - 11 | 1336 | 93 | |
XM_017006222.1 | XP_016861711.1 | 11 - 12 | 1384 | 93 | |
GenBank mRNA | AK310249.1 | - | 10 - 11 | 1183 | 93 |