Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
Chromosome Location
Chr.3: 96814581 - 97761532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX099374.1 - 2 - 3 412 64

View Full Product Details