The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282959.1 | NP_001269888.1 | 13 - 14 | 1977 | 84 |
NM_001282960.1 | NP_001269889.1 | 13 - 14 | 1971 | 84 | |
NM_018237.3 | NP_060707.2 | 14 - 15 | 2016 | 84 | |
GenBank mRNA | AF465616.1 | - | 14 - 15 | 1946 | 84 |
AK000741.1 | - | 9 - 10 | 1330 | 84 | |
AK001701.1 | - | 14 - 15 | 1924 | 84 | |
AK023438.1 | - | 14 - 15 | 1946 | 84 | |
AK122871.1 | - | 12 - 13 | 1791 | 84 | |
AK292649.1 | - | 14 - 15 | 1921 | 84 | |
AK298004.1 | - | 13 - 14 | 1896 | 84 | |
AK299307.1 | - | 13 - 14 | 1876 | 84 | |
AK307550.1 | - | 9 - 10 | 1336 | 84 | |
AK307671.1 | - | 12 - 13 | 1840 | 84 | |
AK307704.1 | - | 14 - 15 | 1921 | 84 | |
AY249140.1 | - | 14 - 15 | 1924 | 84 | |
BC026036.2 | - | 13 - 14 | 1878 | 84 | |
BC089420.1 | - | 13 - 14 | 1838 | 84 | |
BC108682.1 | - | 14 - 15 | 1894 | 84 | |
BC130626.1 | - | 14 - 15 | 1905 | 84 | |
BC132725.1 | - | 14 - 15 | 1905 | 84 | |
BX647943.1 | - | 12 - 13 | 1751 | 84 |