The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018237.3 | NP_060707.2 | 3 - 4 | 430 | 85 |
NR_104262.1 | 3 - 4 | 436 | 85 | ||
GenBank mRNA | AF465616.1 | - | 3 - 4 | 360 | 85 |
AK001701.1 | - | 3 - 4 | 338 | 85 | |
AK023438.1 | - | 3 - 4 | 360 | 85 | |
AK292649.1 | - | 3 - 4 | 335 | 85 | |
AY249140.1 | - | 3 - 4 | 338 | 85 | |
BC026036.2 | - | 2 - 3 | 292 | 85 | |
BC089420.1 | - | 2 - 3 | 252 | 85 | |
BC108682.1 | - | 3 - 4 | 308 | 85 | |
BC130626.1 | - | 3 - 4 | 319 | 85 | |
BC132725.1 | - | 3 - 4 | 319 | 85 | |
BC144118.1 | - | 3 - 4 | 325 | 85 | |
BM786237.1 | - | 3 - 4 | 342 | 85 | |
DA511293.1 | - | 3 - 4 | 430 | 85 |