The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300780.1 | NP_001287709.1 | 12 - 13 | 2025 | 68 |
NM_020796.4 | NP_065847.1 | 12 - 13 | 2025 | 68 | |
XM_005272042.3 | XP_005272099.2 | 12 - 13 | 1589 | 68 | |
XM_006714663.1 | XP_006714726.1 | 12 - 13 | 1924 | 68 | |
XM_017009675.1 | XP_016865164.1 | 12 - 13 | 1329 | 68 | |
XM_017009676.1 | XP_016865165.1 | 7 - 8 | 3876 | 68 | |
GenBank mRNA | AB037789.1 | - | 12 - 13 | 1494 | 68 |
AF279656.1 | - | 12 - 13 | 1902 | 68 | |
AK027501.1 | - | 3 - 4 | 336 | 68 | |
AK027867.1 | - | 10 - 11 | 1150 | 68 | |
AK315143.1 | - | 12 - 13 | 1444 | 68 | |
BC032619.1 | - | 12 - 13 | 2036 | 68 |