Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
Chromosome Location
Chr.6: 83552885 - 83709408 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242792.1 NP_001229721.1 25 - 26 2648 96
NM_001242793.1 NP_001229722.1 23 - 24 2357 96
NM_001242794.1 NP_001229723.1 15 - 16 1526 96
NM_001256717.1 NP_001243646.1 25 - 26 2715 96
NM_001256718.1 NP_001243647.1 24 - 25 2343 96
NM_014841.2 NP_055656.1 25 - 26 2651 96
XM_005248770.4 XP_005248827.1 25 - 26 2490 96
XM_006715615.1 XP_006715678.1 25 - 26 2442 96
XM_011536265.1 XP_011534567.1 25 - 26 2449 96
XM_011536266.1 XP_011534568.1 25 - 26 2561 96
XM_011536269.1 XP_011534571.1 25 - 26 2427 96
XM_011536271.1 XP_011534573.1 24 - 25 2400 96
XM_011536273.2 XP_011534575.1 24 - 25 2364 96
XM_011536275.2 XP_011534577.1 24 - 25 2358 96
XM_011536276.2 XP_011534578.1 24 - 25 2334 96
XM_011536277.1 XP_011534579.1 27 - 28 2730 96
XM_017011554.1 XP_016867043.1 24 - 25 2600 96
XM_017011555.1 XP_016867044.1 24 - 25 2484 96
XM_017011556.1 XP_016867045.1 24 - 25 2416 96
XM_017011557.1 XP_016867046.1 25 - 26 2604 96
XM_017011558.1 XP_016867047.1 25 - 26 2434 96
XM_017011559.1 XP_016867048.1 25 - 26 2533 96
XM_017011560.1 XP_016867049.1 25 - 26 2475 96
XM_017011561.1 XP_016867050.1 25 - 26 2546 96
XM_017011562.1 XP_016867051.1 24 - 25 2604 96
XM_017011563.1 XP_016867052.1 23 - 24 2598 96
XM_017011564.1 XP_016867053.1 23 - 24 2469 96
XM_017011565.1 XP_016867054.1 23 - 24 2527 96
XM_017011566.1 XP_016867055.1 24 - 25 2577 96
XM_017011567.1 XP_016867056.1 24 - 25 2380 96
XM_017011568.1 XP_016867057.1 23 - 24 2571 96
XM_017011569.1 XP_016867058.1 24 - 25 2562 96
XM_017011570.1 XP_016867059.1 24 - 25 2541 96
XM_017011571.1 XP_016867060.1 24 - 25 2412 96
XM_017011572.1 XP_016867061.1 24 - 25 2371 96
XM_017011573.1 XP_016867062.1 24 - 25 2470 96
XM_017011574.1 XP_016867063.1 25 - 26 2619 96
XM_017011575.1 XP_016867064.1 24 - 25 2535 96
XM_017011576.1 XP_016867065.1 24 - 25 2338 96
XM_017011577.1 XP_016867066.1 24 - 25 2464 96
XM_017011578.1 XP_016867067.1 24 - 25 2477 96
XM_017011579.1 XP_016867068.1 24 - 25 2520 96
XM_017011580.1 XP_016867069.1 24 - 25 2511 96
XM_017011581.1 XP_016867070.1 24 - 25 2341 96
XM_017011582.1 XP_016867071.1 24 - 25 2370 96
XM_017011583.1 XP_016867072.1 24 - 25 2483 96
XM_017011584.1 XP_016867073.1 24 - 25 2326 96
XM_017011585.1 XP_016867074.1 24 - 25 2367 96
XM_017011586.1 XP_016867075.1 24 - 25 2319 96
XM_017011587.1 XP_016867076.1 24 - 25 2425 96
XM_017011588.1 XP_016867077.1 24 - 25 2438 96
XM_017011589.1 XP_016867078.1 23 - 24 2469 96
XM_017011590.1 XP_016867079.1 23 - 24 2298 96
GenBank mRNA AB014556.1 - 25 - 26 2577 96
AK289582.1 - 25 - 26 2619 96
AK294556.1 - 18 - 19 1609 96
AK294898.1 - 24 - 25 2327 96
BC060818.1 - 15 - 16 1510 96

View Full Product Details