The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276503.1 | NP_001263432.1 | 1 - 2 | 133 | 62 |
NM_001276506.1 | NP_001263435.1 | 1 - 2 | 133 | 62 | |
NM_003002.3 | NP_002993.1 | 1 - 2 | 133 | 62 | |
NR_077060.1 | 1 - 2 | 133 | 62 | ||
GenBank mRNA | AB006202.1 | - | 1 - 2 | 60 | 62 |
AK075360.1 | - | 1 - 2 | 59 | 62 | |
BC005263.1 | - | 1 - 2 | 77 | 62 | |
BC009574.1 | - | 1 - 2 | 58 | 62 | |
BC012603.1 | - | 1 - 2 | 62 | 62 | |
BC015188.1 | - | 1 - 2 | 84 | 62 | |
BC015992.1 | - | 1 - 2 | 80 | 62 | |
BC022350.1 | - | 1 - 2 | 72 | 62 | |
BC070307.1 | - | 1 - 2 | 58 | 62 | |
BC071755.1 | - | 1 - 2 | 76 | 62 | |
BC071756.1 | - | 1 - 2 | 58 | 62 | |
BG492859.1 | - | 1 - 2 | 60 | 62 | |
BQ932668.1 | - | 1 - 2 | 141 | 62 | |
BT007238.1 | - | 1 - 2 | 49 | 62 | |
CD105572.1 | - | 1 - 2 | 73 | 62 | |
CR456932.1 | - | 1 - 2 | 49 | 62 | |
KU178325.1 | - | 1 - 2 | 49 | 62 | |
KU178326.1 | - | 1 - 2 | 49 | 62 |