The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134363.2 | NP_001127835.2 | 13 - 14 | 3625 | 81 |
XM_011539697.2 | XP_011537999.1 | 13 - 14 | 3463 | 81 | |
XM_017016103.1 | XP_016871592.1 | 13 - 14 | 4883 | 81 | |
XM_017016104.1 | XP_016871593.1 | 13 - 14 | 7099 | 81 | |
GenBank mRNA | AK302802.1 | - | 3 - 4 | 442 | 81 |
BX648563.1 | - | 3 - 4 | 490 | 81 | |
EU822950.1 | - | 13 - 14 | 3625 | 81 |