Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
Chromosome Location
Chr.13: 94437304 - 94550265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129889.2 NP_001123361.1 2 - 3 1021 120
NM_001322184.1 NP_001309113.1 3 - 4 597 120
NM_001322185.1 NP_001309114.1 2 - 3 543 120
NM_001322186.1 NP_001309115.1 4 - 5 812 120
NM_001922.4 NP_001913.2 2 - 3 1021 120
XM_011521049.1 XP_011519351.1 4 - 5 1645 120
XM_017020401.1 XP_016875890.1 2 - 3 1024 120
GenBank mRNA AK293115.1 - 2 - 3 804 120
BC028311.1 - 2 - 3 868 120
D17547.1 - 2 - 3 1008 120
DQ902581.1 - 2 - 3 886 120
L18967.1 - 2 - 3 978 120
S69231.1 - 2 - 3 943 120

View Full Product Details