The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129889.2 | NP_001123361.1 | 9 - 10 | 1902 | 64 |
NM_001322182.1 | NP_001309111.1 | 6 - 7 | 1025 | 64 | |
NM_001322183.1 | NP_001309112.1 | 7 - 8 | 1079 | 64 | |
NM_001322184.1 | NP_001309113.1 | 8 - 9 | 1379 | 64 | |
NM_001322185.1 | NP_001309114.1 | 7 - 8 | 1325 | 64 | |
NM_001322186.1 | NP_001309115.1 | 9 - 10 | 1594 | 64 | |
NM_001922.4 | NP_001913.2 | 7 - 8 | 1803 | 64 | |
XM_011521049.1 | XP_011519351.1 | 9 - 10 | 2427 | 64 | |
XM_017020401.1 | XP_016875890.1 | 9 - 10 | 1935 | 64 | |
GenBank mRNA | AJ132932.1 | - | 1 - 2 | 197 | 64 |
AK293115.1 | - | 7 - 8 | 1586 | 64 | |
BC028311.1 | - | 7 - 8 | 1650 | 64 | |
D17547.1 | - | 7 - 8 | 1790 | 64 | |
DQ902581.1 | - | 9 - 10 | 1767 | 64 | |
L18967.1 | - | 7 - 8 | 1760 | 64 | |
S69231.1 | - | 7 - 8 | 1725 | 64 |