The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000821.6 | NP_000812.2 | 10 - 11 | 1541 | 94 |
NM_001142269.3 | NP_001135741.1 | 9 - 10 | 1370 | 94 | |
XM_005264259.4 | XP_005264316.1 | 10 - 11 | 1500 | 94 | |
XM_011532764.2 | XP_011531066.1 | 9 - 10 | 1336 | 94 | |
XM_011532765.2 | XP_011531067.1 | 8 - 9 | 1175 | 94 | |
XM_017003803.1 | XP_016859292.1 | 9 - 10 | 1363 | 94 | |
GenBank mRNA | AK130139.1 | - | 1 - 2 | 133 | 94 |
AK222827.1 | - | 10 - 11 | 1501 | 94 | |
AK225663.1 | - | 10 - 11 | 1500 | 94 | |
AK297397.1 | - | 9 - 10 | 1310 | 94 | |
AK313126.1 | - | 10 - 11 | 1509 | 94 | |
BC013979.2 | - | 10 - 11 | 1463 | 94 | |
EU847509.1 | - | 10 - 11 | 1438 | 94 | |
L17128.1 | - | 10 - 11 | 1590 | 94 | |
M81592.1 | - | 10 - 11 | 1521 | 94 |