The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000821.6 | NP_000812.2 | 11 - 12 | 1712 | 83 |
NM_001142269.3 | NP_001135741.1 | 10 - 11 | 1541 | 83 | |
XM_005264259.4 | XP_005264316.1 | 11 - 12 | 1671 | 83 | |
XM_011532764.2 | XP_011531066.1 | 10 - 11 | 1507 | 83 | |
XM_011532765.2 | XP_011531067.1 | 9 - 10 | 1346 | 83 | |
XM_017003803.1 | XP_016859292.1 | 10 - 11 | 1534 | 83 | |
GenBank mRNA | AK130139.1 | - | 2 - 3 | 304 | 83 |
AK222827.1 | - | 11 - 12 | 1672 | 83 | |
AK225663.1 | - | 11 - 12 | 1671 | 83 | |
AK297397.1 | - | 10 - 11 | 1481 | 83 | |
AK298982.1 | - | 10 - 11 | 1366 | 83 | |
AK313126.1 | - | 11 - 12 | 1680 | 83 | |
BC013979.2 | - | 11 - 12 | 1634 | 83 | |
EU847509.1 | - | 11 - 12 | 1609 | 83 | |
L17128.1 | - | 11 - 12 | 1761 | 83 | |
M81592.1 | - | 11 - 12 | 1692 | 83 |