The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024074.2 | NP_076979.1 | 3 - 4 | 559 | 72 |
GenBank mRNA | AK025981.1 | - | 3 - 4 | 559 | 72 |
AK292764.1 | - | 3 - 4 | 544 | 72 | |
BC001195.1 | - | 3 - 4 | 475 | 72 |